Assay Details
Gene Symbol: LOC694289 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001083221.4 | XP_001083221.2 | 3065 | 68 | ||
XM_015148704.2 | XP_015004190.2 | 3068 | 68 | |||
XM_015148705.2 | XP_015004191.2 | 3023 | 68 | |||
XM_015148706.2 | XP_015004192.2 | 3020 | 68 | |||
XM_015148707.2 | XP_015004193.1 | 639 | 68 | |||
XM_015148708.2 | XP_015004194.1 | 506 | 68 | |||
XM_028828531.1 | XP_028684364.1 | 921 | 68 | |||
XM_028828532.1 | XP_028684365.1 | 957 | 68 | |||
XM_028828533.1 | XP_028684366.1 | 876 | 68 | |||
XM_028828534.1 | XP_028684367.1 | 912 | 68 | |||
XM_028828535.1 | XP_028684368.1 | 460 | 68 | |||
XM_028828536.1 | XP_028684369.1 | 1555 | 68 | |||
XM_028828537.1 | XP_028684370.1 | 437 | 68 | |||
XM_028828538.1 | XP_028684371.1 | 559 | 68 | |||
XM_028828539.1 | XP_028684372.1 | 617 | 68 | |||
XM_028828540.1 | XP_028684373.1 | 449 | 68 | |||
GenBank mRNA | JV043979.1 | - | 579 | 68 | ||
JV043980.1 | - | 579 | 68 | |||
JV600239.1 | - | 510 | 68 | |||
JV600240.1 | - | 513 | 68 | |||
JV712359.1 | - | 789 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
