Assay Details
Gene Symbol: HPF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260760.2 | 90 | 97 | |||
XM_015139454.2 | XP_014994940.1 | 129 | 97 | |||
GenBank mRNA | JU323133.1 | - | 96 | 97 | ||
JV045476.1 | - | 90 | 97 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
