Assay Details
| Gene Symbol: NSMCE2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015145977.2 | XP_015001463.1 | 283 | 80 | ||
| XM_015145978.2 | XP_015001464.1 | 392 | 80 | |||
| XM_015145979.2 | XP_015001465.1 | 312 | 80 | |||
| XM_015145980.2 | XP_015001466.1 | 263 | 80 | |||
| XM_015145982.2 | XP_015001468.1 | 284 | 80 | |||
| XM_015145983.2 | XP_015001469.2 | 285 | 80 | |||
| XM_028852907.1 | XP_028708740.1 | 370 | 80 | |||
| GenBank mRNA | JU334110.1 | - | 217 | 80 | ||
| JU475254.1 | - | 330 | 80 | |||
| JV058782.1 | - | 360 | 80 | |||
More Information
Set Membership: |
|

NSMCE2
Genomic Map