Assay Details
| Gene Symbol: SNRPB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260894.1 | NP_001247823.1 | 516 | 94 | ||
| XM_015149346.2 | XP_015004832.1 | 785 | 94 | |||
| GenBank mRNA | JU321746.1 | - | 418 | 94 | ||
| JU473447.1 | - | 466 | 94 | |||
| JV044849.1 | - | 516 | 94 | |||
| JV452520.1 | - | 465 | 94 | |||
| JV490286.1 | - | 478 | 94 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

SNRPB2
Genomic Map