Assay Details
Gene Symbol: ADTRP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193850.2 | NP_001180779.1 | 747 | 60 | ||
XM_015135467.2 | XP_014990953.1 | 1072 | 60 | |||
XM_015135468.2 | XP_014990954.1 | 956 | 60 | |||
GenBank mRNA | JU323455.1 | - | 747 | 60 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() ![]() |
