Assay Details
Gene Symbol: NHLRC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001087923.4 | XP_001087923.1 | 910 | 75 | ||
GenBank mRNA | JU334445.1 | - | 887 | 75 | ||
JU334446.1 | - | 887 | 75 | |||
JV047020.1 | - | 864 | 75 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
