Assay Details
| Gene Symbol: MCRIP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001193773.2 | NP_001180702.1 | 476 | 75 | ||
| XM_015125165.2 | XP_014980651.1 | 477 | 75 | |||
| GenBank mRNA | JU325823.1 | - | 136 | 75 | ||
| JV566185.1 | - | 450 | 75 | |||
| JV635710.1 | - | 431 | 75 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

MCRIP2
Genomic Map