Assay Details
Gene Symbol: ADRM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260529.1 | NP_001247458.1 | 419 | 92 | ||
XM_015148636.2 | XP_015004122.1 | 471 | 92 | |||
XM_028827217.1 | XP_028683050.1 | 421 | 92 | |||
GenBank mRNA | JU324550.1 | - | 364 | 92 | ||
JV043777.1 | - | 419 | 92 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() ![]() |
