Assay Details
| Gene Symbol: SPINK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015138626.1 | XP_014994112.1 | 383 | 72 | ||
| GenBank mRNA | JU324943.1 | - | 62 | 72 | ||
| JU476925.1 | - | 219 | 72 | |||
More Information
Set Membership: |
|

SPINK2
Genomic Map