Assay Details
| Gene Symbol: EIF4E | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266219.2 | NP_001253148.1 | 406 | 90 | ||
| GenBank mRNA | JU320282.1 | - | 418 | 90 | ||
| JU470709.1 | - | 413 | 90 | |||
| JV044801.1 | - | 409 | 90 | |||
| JV675335.1 | - | 395 | 90 | |||
More Information
Set Membership: |
|

EIF4E
Genomic Map