Assay Details
Gene Symbol: LOC114675313 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028842554.1 | XP_028698387.1 | 359 | 141 |
Gene Symbol: CNN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261809.1 | NP_001248738.1 | 606 | 141 | ||
XM_015145070.2 | XP_015000556.1 | 445 | 141 | |||
GenBank mRNA | JU319626.1 | - | 509 | 141 | ||
JU472900.1 | - | 549 | 141 | |||
JV044008.1 | - | 606 | 141 | |||
JV451657.1 | - | 624 | 141 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() |
