Assay Details
| Gene Symbol: PRDX6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266085.1 | NP_001253014.1 | 312 | 109 | ||
| GenBank mRNA | JU327091.1 | - | 153 | 109 | ||
| JU472813.1 | - | 312 | 109 | |||
| JV048785.1 | - | 312 | 109 | |||
More Information
Set Membership: |
|

PRDX6
Genomic Map