Assay Details
Gene Symbol: GPBP1L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261345.1 | NP_001248274.1 | 1997 | 93 | ||
XM_015138369.2 | XP_014993855.1 | 2196 | 93 | |||
XM_015138379.2 | XP_014993865.1 | 2177 | 93 | |||
XM_015138385.2 | XP_014993871.1 | 2095 | 93 | |||
XM_015138389.2 | XP_014993875.1 | 2007 | 93 | |||
GenBank mRNA | JU321138.1 | - | 1954 | 93 | ||
JU471282.1 | - | 1997 | 93 | |||
JU471283.1 | - | 1997 | 93 | |||
JU471284.1 | - | 1997 | 93 | |||
JV043934.1 | - | 1997 | 93 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() ![]() |
