Assay Details
Gene Symbol: PRDX3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001265826.2 | NP_001252755.1 | 183 | 101 | ||
GenBank mRNA | JU479186.1 | - | 110 | 101 | ||
JV600249.1 | - | 183 | 101 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
