Assay Details
Gene Symbol: RHEB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261182.1 | NP_001248111.1 | 167 | 103 | ||
XM_028845048.1 | XP_028700881.1 | 122 | 103 | |||
GenBank mRNA | JU320547.1 | - | 352 | 103 | ||
JU470465.1 | - | 348 | 103 | |||
JV047844.1 | - | 167 | 103 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
