Assay Details
Gene Symbol: MED27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266508.1 | NP_001253437.1 | 214 | 63 | ||
XM_015116518.2 | XP_014972004.2 | 264 | 63 | |||
XM_015116519.2 | XP_014972005.1 | 264 | 63 | |||
GenBank mRNA | JU337177.1 | - | 209 | 63 | ||
JU477905.1 | - | 213 | 63 | |||
JV045727.1 | - | 214 | 63 | |||
JV600556.1 | - | 219 | 63 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
