Assay Details
| Gene Symbol: CEP19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260983.1 | NP_001247912.1 | 502 | 113 | ||
| XM_015132576.2 | XP_014988062.1 | 331 | 113 | |||
| GenBank mRNA | JU336809.1 | - | 502 | 113 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

CEP19
Genomic Map