Assay Details
Gene Symbol: CDC5L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261272.1 | NP_001248201.1 | 448 | 66 | ||
GenBank mRNA | JU337371.1 | - | 487 | 66 | ||
JU478068.1 | - | 487 | 66 | |||
JV044138.1 | - | 448 | 66 | |||
JV703472.1 | - | 635 | 66 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() ![]() |
