Assay Details
Gene Symbol: TSPAN5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194290.2 | NP_001181219.1 | 981 | 95 | ||
XM_001104834.4 | XP_001104834.1 | 1074 | 95 | |||
XM_015138828.2 | XP_014994314.1 | 927 | 95 | |||
GenBank mRNA | JU332404.1 | - | 993 | 95 | ||
JU473865.1 | - | 743 | 95 | |||
JV043788.1 | - | 743 | 95 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() |
