Assay Details
| Gene Symbol: SLCO1B1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028829310.1 | XP_028685143.1 | 541 | 144 | ||
| XM_028829311.1 | XP_028685144.1 | 541 | 144 | |||
| GenBank mRNA | JV705369.1 | - | 217 | 144 | ||
More Information
Set Membership: |
|

SLCO1B1
Genomic Map