Assay Details
Gene Symbol: ERG28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194135.3 | NP_001181064.1 | 580 | 88 | ||
GenBank mRNA | JU332721.1 | - | 154 | 88 | ||
JU470961.1 | - | 288 | 88 | |||
JV044875.1 | - | 272 | 88 | |||
JV637984.1 | - | 392 | 88 |
