Assay Details
Gene Symbol: ERG28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194135.3 | NP_001181064.1 | 661 | 64 | ||
GenBank mRNA | JU332721.1 | - | 235 | 64 | ||
JU470961.1 | - | 369 | 64 | |||
JV044875.1 | - | 353 | 64 | |||
JV637984.1 | - | 473 | 64 |

More Information
Set Membership: |
![]() ![]() |
