Assay Details
Gene Symbol: ERG28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194135.3 | NP_001181064.1 | 785 | 71 | ||
GenBank mRNA | JU332721.1 | - | 359 | 71 | ||
JU470961.1 | - | 493 | 71 | |||
JV044875.1 | - | 477 | 71 | |||
JV637984.1 | - | 597 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() |
