Assay Details
| Gene Symbol: SLAIN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266145.1 | NP_001253074.1 | 1752 | 81 | ||
| XM_015138322.2 | XP_014993808.1 | 2593 | 81 | |||
| GenBank mRNA | JU328296.1 | - | 1357 | 81 | ||
| JU471627.1 | - | 1806 | 81 | |||
| JV044093.1 | - | 1752 | 81 | |||
| JV639139.1 | - | 1868 | 81 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

SLAIN2
Genomic Map