Assay Details
| Gene Symbol: SPOCK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266535.1 | NP_001253464.1 | 825 | 56 | ||
| XM_015140905.2 | XP_014996391.1 | 877 | 56 | |||
| GenBank mRNA | JU321723.1 | - | 773 | 56 | ||
| JU321724.1 | - | 773 | 56 | |||
| JU336062.1 | - | 782 | 56 | |||
| JV048025.1 | - | 825 | 56 | |||
| JV591740.1 | - | 842 | 56 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

SPOCK1
Genomic Map