Assay Details
| Gene Symbol: CFAP91 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194476.2 | NP_001181405.2 | 2224 | 111 | ||
| XM_028843343.1 | XP_028699176.1 | 2230 | 111 | |||
| XM_028843344.1 | XP_028699177.1 | 1828 | 111 | |||
| GenBank mRNA | JV597550.1 | - | 650 | 111 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

CFAP91
Genomic Map