Assay Details
Gene Symbol: RAMAC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001265849.1 | NP_001252778.1 | 132 | 64 | ||
GenBank mRNA | JU321796.1 | - | 132 | 64 | ||
JU472742.1 | - | 135 | 64 | |||
JV043700.1 | - | 132 | 64 | |||
JV638339.1 | - | 137 | 64 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001265849.1 transcripts.
Set Membership: |
![]() ![]() |
