Assay Details
Gene Symbol: RAMAC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001265849.1 | NP_001252778.1 | 172 | 82 | ||
GenBank mRNA | JU321796.1 | - | 172 | 82 | ||
JU472742.1 | - | 175 | 82 | |||
JV043700.1 | - | 172 | 82 | |||
JV638339.1 | - | 177 | 82 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001265849.1 transcripts.
Set Membership: |
![]() ![]() |
