Assay Details
| Gene Symbol: IFI6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001048170.1 | NP_001041635.1 | 408 | 106 | ||
| XM_015132328.2 | XP_014987814.1 | 525 | 106 | |||
| XM_028845584.1 | XP_028701417.1 | 355 | 106 | |||
| GenBank mRNA | JU323453.1 | - | 404 | 106 | ||
| JU332230.1 | - | 404 | 106 | |||
| JU332231.1 | - | 404 | 106 | |||
| JU473079.1 | - | 426 | 106 | |||
| JU473080.1 | - | 426 | 106 | |||
| JU473081.1 | - | 426 | 106 | |||
| JU473082.1 | - | 426 | 106 | |||
| JV046642.1 | - | 431 | 106 | |||
| JV046643.1 | - | 431 | 106 | |||
| JV046644.1 | - | 431 | 106 | |||
| JV046645.1 | - | 431 | 106 | |||
| JV046646.1 | - | 431 | 106 | |||
| JV046647.1 | - | 431 | 106 | |||
More Information
Set Membership: |
|

IFI6
Genomic Map