Assay Details
Gene Symbol: DRAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194542.1 | NP_001181471.1 | 358 | 88 | ||
XM_028832767.1 | XP_028688600.1 | 384 | 88 | |||
GenBank mRNA | JU322299.1 | - | 204 | 88 | ||
JU472469.1 | - | 126 | 88 | |||
JV045317.1 | - | 223 | 88 | |||
JV602177.1 | - | 322 | 88 | |||
JV636505.1 | - | 275 | 88 | |||
JV668253.1 | - | 345 | 88 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
