Assay Details
Gene Symbol: CARD19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260938.1 | NP_001247867.1 | 218 | 91 | ||
XM_015117248.2 | XP_014972734.1 | 367 | 91 | |||
XM_015117249.2 | XP_014972735.1 | 367 | 91 | |||
GenBank mRNA | JU321018.1 | - | 285 | 91 | ||
JU472062.1 | - | 218 | 91 | |||
JV637639.1 | - | 314 | 91 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() |
