Assay Details
Gene Symbol: IDH3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266087.2 | NP_001253016.1 | 924 | 71 | ||
XM_015149351.2 | XP_015004837.2 | 1304 | 71 | |||
XM_015149352.2 | XP_015004838.2 | 1300 | 71 | |||
XM_015149354.2 | XP_015004840.1 | 1346 | 71 | |||
XM_028827410.1 | XP_028683243.1 | 1300 | 71 | |||
XM_028827411.1 | XP_028683244.1 | 1300 | 71 | |||
XM_028827412.1 | XP_028683245.1 | 1346 | 71 | |||
XM_028827413.1 | XP_028683246.1 | 1346 | 71 | |||
XM_028827414.1 | XP_028683247.1 | 1346 | 71 | |||
GenBank mRNA | JU321303.1 | - | 919 | 71 | ||
JU322233.1 | - | 919 | 71 | |||
JU475429.1 | - | 924 | 71 | |||
JV529301.1 | - | 920 | 71 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
