Assay Details
| Gene Symbol: GZMB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001114420.4 | XP_001114420.3 | 720 | 126 | ||
| GenBank mRNA | JU475347.1 | - | 657 | 126 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

GZMB| Gene Symbol: GZMB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001114420.4 | XP_001114420.3 | 720 | 126 | ||
| GenBank mRNA | JU475347.1 | - | 657 | 126 | ||
Set Membership: |
|