Assay Details
Gene Symbol: BUD23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266959.1 | NP_001253888.1 | 114 | 89 | ||
GenBank mRNA | JU332790.1 | - | 120 | 89 | ||
JU332791.1 | - | 120 | 89 | |||
JU332792.1 | - | 120 | 89 | |||
JU474210.1 | - | 120 | 89 | |||
JV043191.1 | - | 114 | 89 | |||
JV706194.1 | - | 136 | 89 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
