Assay Details
Gene Symbol: BUD23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266959.1 | NP_001253888.1 | 665 | 98 | ||
XM_028845071.1 | XP_028700904.1 | 791 | 98 | |||
GenBank mRNA | JU332790.1 | - | 671 | 98 | ||
JU332791.1 | - | 671 | 98 | |||
JU332792.1 | - | 671 | 98 | |||
JU474210.1 | - | 671 | 98 | |||
JV043191.1 | - | 665 | 98 | |||
JV706194.1 | - | 687 | 98 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
