Assay Details
Gene Symbol: CD99 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015126881.2 | XP_014982367.1 | 442 | 112 | ||
XM_015126882.2 | XP_014982368.1 | 442 | 112 | |||
XM_015126884.2 | XP_014982370.1 | 360 | 112 | |||
XM_015126885.2 | XP_014982371.1 | 360 | 112 | |||
XM_015126887.2 | XP_014982373.1 | 442 | 112 | |||
GenBank mRNA | JU473061.1 | - | 338 | 112 | ||
JV043984.1 | - | 322 | 112 | |||
JV043985.1 | - | 370 | 112 | |||
JV651792.1 | - | 340 | 112 |

More Information
Set Membership: |
![]() ![]() ![]() |
