Assay Details
| Gene Symbol: C1H1orf21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260863.1 | NP_001247792.1 | 78 | 71 | ||
| GenBank mRNA | JU319728.1 | - | 301 | 71 | ||
| JU477354.1 | - | 115 | 71 | |||
| JV048336.1 | - | 78 | 71 | |||
| JV566003.1 | - | 328 | 71 | |||
| JV601878.1 | - | 361 | 71 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001260863.1 transcripts.
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

C1H1orf21
Genomic Map