Assay Details
Gene Symbol: MAMU-E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001114966.1 | NP_001108438.1 | 1029 | 69 | ||
GenBank mRNA | EF372279.1 | - | 1029 | 69 | ||
EF611169.1 | - | 1026 | 69 | |||
EF611170.1 | - | 1029 | 69 | |||
EF611171.1 | - | 1029 | 69 | |||
EU109710.1 | - | 1026 | 69 | |||
EU109711.1 | - | 1026 | 69 | |||
EU305658.1 | - | 1029 | 69 | |||
EU305660.1 | - | 1029 | 69 | |||
EU934770.1 | - | 1029 | 69 | |||
FJ544410.1 | - | 1029 | 69 | |||
GQ471889.1 | - | 1029 | 69 | |||
GQ471890.1 | - | 1029 | 69 | |||
HM106999.1 | - | 1029 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
