Assay Details
| Gene Symbol: TRAPPC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266767.2 | NP_001253696.1 | 328 | 82 | ||
| XM_015135124.2 | XP_014990610.1 | 646 | 82 | |||
| XM_015135130.2 | XP_014990616.1 | 218 | 82 | |||
| GenBank mRNA | JU320973.1 | - | 334 | 82 | ||
| JU473488.1 | - | 335 | 82 | |||
| JV045481.1 | - | 328 | 82 | |||
| JV600882.1 | - | 335 | 82 | |||
More Information
Set Membership: |
|

TRAPPC3
Genomic Map