Assay Details
| Gene Symbol: ANAPC13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001190866.1 | NP_001177795.1 | 164 | 66 | ||
| XM_015132065.2 | XP_014987551.1 | 287 | 66 | |||
| GenBank mRNA | CB311140.1 | - | 167 | 66 | ||
| CN641675.1 | - | 216 | 66 | |||
| JV045584.1 | - | 159 | 66 | |||
| Gene Symbol: CEP63 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | JV637096.1 | - | 1026 | 66 | ||
More Information
Set Membership: |
|

ANAPC13
Genomic Map