Assay Details
| Gene Symbol: SNRPB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257716.1 | NP_001244645.1 | 446 | 131 | ||
| XM_015149349.2 | XP_015004835.1 | 605 | 131 | |||
| GenBank mRNA | JU320714.1 | - | 446 | 131 | ||
| JU323736.1 | - | 446 | 131 | |||
| JU470796.1 | - | 689 | 131 | |||
| JU476906.1 | - | 689 | 131 | |||
| JV600898.1 | - | 484 | 131 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

SNRPB
Genomic Map