Assay Details
| Gene Symbol: COX7C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | JU480496.1 | - | 295 | 102 | ||
| Gene Symbol: LOC106998836 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028849562.1 | XP_028705395.1 | 355 | 102 | ||
| GenBank mRNA | JV606203.1 | - | 308 | 102 | ||
More Information
Set Membership: |
|

COX7C
Genomic Map