Assay Details
| Gene Symbol: RAB4A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257551.1 | NP_001244480.1 | 226 | 83 | ||
| XM_028826814.1 | XP_028682647.1 | 193 | 83 | |||
| GenBank mRNA | JU335734.1 | - | 226 | 83 | ||
| JU476623.1 | - | 190 | 83 | |||
| JV047812.1 | - | 158 | 83 | |||
| JV720273.1 | - | 238 | 83 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_028826814.1 transcripts.
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

RAB4A
Genomic Map