Assay Details
| Gene Symbol: MRPL47 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001265881.1 | NP_001252810.1 | 313 | 102 | ||
| XM_015131551.2 | XP_014987037.1 | 538 | 102 | |||
| XM_028843306.1 | XP_028699139.1 | 304 | 102 | |||
| XM_028843307.1 | XP_028699140.1 | 208 | 102 | |||
| GenBank mRNA | JU321052.1 | - | 323 | 102 | ||
| JU476490.1 | - | 322 | 102 | |||
| JV045103.1 | - | 313 | 102 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_028843307.1 transcripts.
Set Membership: |
|

MRPL47
Genomic Map