Assay Details
Gene Symbol: LOC719235 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028839442.1 | XP_028695275.1 | 625 | 99 | ||
GenBank mRNA | JU496642.1 | - | 353 | 99 | ||
JU496643.1 | - | 353 | 99 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() ![]() |
