Assay Details
| Gene Symbol: MRE11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015115484.2 | XP_014970970.2 | 2153 | 82 | ||
| XM_015115486.2 | XP_014970972.2 | 2150 | 82 | |||
| XM_028833912.1 | XP_028689745.1 | 1988 | 82 | |||
| XM_028833913.1 | XP_028689746.1 | 1568 | 82 | |||
| GenBank mRNA | JU334579.1 | - | 2016 | 82 | ||
| JV715422.1 | - | 1931 | 82 | |||
More Information
Set Membership: |
|

MRE11
Genomic Map