Assay Details
| Gene Symbol: BRI3BP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002798836.3 | XP_002798882.3 | 467 | 64 | ||
| GenBank mRNA | JU474375.1 | - | 386 | 64 | ||
| JV642776.1 | - | 324 | 64 | |||
More Information
Set Membership: |
|

BRI3BP
Genomic Map