Assay Details
| Gene Symbol: RPL32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015130125.2 | XP_014985611.1 | 478 | 147 | ||
| GenBank mRNA | JU470751.1 | - | 489 | 147 | ||
| JV635490.1 | - | 412 | 147 | |||
More Information
Set Membership: |
|

RPL32
Genomic Map