Assay Details
Gene Symbol: RAB1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257272.1 | NP_001244201.1 | 646 | 103 | ||
GenBank mRNA | CK230088.1 | - | 337 | 103 | ||
CN642107.1 | - | 479 | 103 | |||
JU320087.1 | - | 625 | 103 | |||
JU320088.1 | - | 625 | 103 | |||
JU476622.1 | - | 594 | 103 | |||
JV047809.1 | - | 1056 | 103 | |||
JV528322.1 | - | 629 | 103 | |||
JV657698.1 | - | 627 | 103 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() ![]() |
