Assay Details
| Gene Symbol: TOB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261597.1 | NP_001248526.1 | 496 | 99 | ||
| GenBank mRNA | JU332595.1 | - | 463 | 99 | ||
| JU332596.1 | - | 463 | 99 | |||
| JU474063.1 | - | 472 | 99 | |||
| JU474064.1 | - | 472 | 99 | |||
| JU474065.1 | - | 472 | 99 | |||
| JV045977.1 | - | 496 | 99 | |||
| JV640601.1 | - | 509 | 99 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001261597.1 transcripts.
Set Membership: |
|

TOB2
Genomic Map